Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRG Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116277
Description
MRG Polyclonal Antibody for Western Blot
C20orf20, also named as MRG/MORF4L-binding protein or Up-regulated in colon cancer 4 (Urcc4), is a 204 amino acid protein, which belongs to the EAF7 family. C20orf20 localizes in the nucleus and is a Component of the NuA4 histone acetyltransferase (HAT) complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histones H4 and H2A. This modification may both alter nucleosome - DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. This complex may be required for the activation of transcriptional programs associated with oncogene and proto-oncogene mediated growth induction, tumor suppressor mediated growth arrest and replicative senescence, apoptosis, and DNA repair. C20orf20 is modified through phosphorylation and acetylation, so the molecular weight of C20orf20 might be 25-30 kDa.
Specifications
| MRG | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9NV56 | |
| MRGBP | |
| KLH conjugated synthetic peptide between 46-75 amino acids from the Central region of human MRGBP. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| MRGBP | |
| 1600027N09Rik; AW060503; C20orf20; C80444; Eaf7; FLJ10914; MRG/MORF4L binding protein; MRG/MORF4L-binding protein; MRG15BP; MRG-binding protein; MRGBP; RGD1308612; Up-regulated in colon cancer 4; Urcc4 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 55257 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction