Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRG15 Antibody (OTI5D2), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272778FR
Description
MRG15 Monoclonal specifically detects MRG15 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MRG15 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
MORF4L1 | |
Recombinant protein fragment corresponding to amino acids 6-77 of human MRG15 (NP_006782) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI5D2 | |
Western Blot, Immunohistochemistry | |
Eaf3, Esa1p-associated factor 3 homolog, HsT17725, MEAF3, MGC10631, MORF-related gene on chromosome 15, MORFRG15, mortality factor 4 like 1, mortality factor 4-like protein 1, MRG15MORF-related gene 15 protein, Protein MSL3-1, S863-6, Transcription factor-like protein MRG15 | |
Mouse | |
Affinity Purified | |
DNA replication Transcription Translation and Splicing | |
10933.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction