Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRG15 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986453
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6453 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6453 Supplier Bioss Supplier No. BS5272R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MRG15 ia a component of the NuA4 histone acetyltransferase (HAT) complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histone H4 and H2A. This modification may both alter nucleosome-DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. This complex may be required for the activation of transcriptional programs associated with oncogene and proto-oncogene mediated growth induction, tumor suppressor mediated growth arrest and replicative senescence, apoptosis and DNA repair.

Specifications

Antigen MRG15
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MORF4L1
Gene Accession No. Q9UBU8
Gene Alias Eaf3; Esa1p-associated factor 3 homolog; FWP006; HSPC008; HSPC061; HsT17725; MEAF3; MGC10631; mKIAA4002; MORF4L1; MORF-related gene 15 protein; MORF-related gene on chromosome 15; MORFRG15; mortality factor 4 like 1; Mortality factor 4-like protein 1; Mrg15; PP368; Protein MSL3-1; S863-6; TEG-189; testis expressed gene 189; Testis-expressed gene 189 protein; Tex189; Transcription factor-like protein MRG15
Gene Symbols MORF4L1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MRG15.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10933
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.