Learn More
Invitrogen™ MRGD Polyclonal Antibody

Description
Antibody detects endogenous levels of total MRGRD.
Specifications
Specifications
| Antigen | MRGD |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | MRGPRD |
| Gene Accession No. | Q8TDS7 |
| Gene Alias | Beta-alanine receptor; G protein-coupled receptor MrgD; Gm499; G-protein coupled receptor TGR7; MAS related GPR family member D; mas-related G protein-coupled MRGD; Mas-related G protein-coupled receptor d; MAS-related gene D; MAS-related GPR, member D; mas-related G-protein coupled receptor member D; MAS-related G-protein coupled receptor, member D; Mgrd; MrgD; MRGD G protein-coupled receptor; MRGPRD; TGR7 |
| Gene Symbols | MRGPRD |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.