Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRGD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533942
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533942 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533942 Supplier Invitrogen™ Supplier No. PA533942
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human (100%) Gorilla, Gibbon, Monkey (94%).

May regulate nociceptor function and/or development, including the sensation or modulation of pain. Functions as a specific membrane receptor for beta-alanine. Beta-alanine at micromolar doses specifically evoked Ca(2+) influx in cells expressing the receptor. Beta-alanine decreases forskolin-stimulated cAMP production in cells expressing the receptor, suggesting that the receptor couples with G-protein G(q) and G(i). Localized at the plasma membrane but internalized into the cytoplasm after treatment with beta-alanine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRGD
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene MRGPRD
Gene Accession No. Q8TDS7
Gene Alias Beta-alanine receptor; G protein-coupled receptor MrgD; Gm499; G-protein coupled receptor TGR7; MAS related GPR family member D; mas-related G protein-coupled MRGD; Mas-related G protein-coupled receptor d; MAS-related gene D; MAS-related GPR, member D; mas-related G-protein coupled receptor member D; MAS-related G-protein coupled receptor, member D; Mgrd; MrgD; MRGD G protein-coupled receptor; MRGPRD; TGR7
Gene Symbols MRGPRD
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from 2nd extracellular domain of human MRGPRD.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116512
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.