Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRGPRE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111236
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111236 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111236 Supplier Invitrogen™ Supplier No. PA5111236
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MMEPREAGQH VGAANGAQED VAFNLII Highest antigen sequence indentity to the following orthologs: Rat - 41%, Mouse - 41%.

Orphan receptor. May regulate nociceptor function and/or development, including the sensation or modulation of pain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRGPRE
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MRGPRE
Gene Accession No. Q86SM8
Gene Alias C130069N09Rik; Ebrt3; Evolutionary breakpoint transcript 3 protein; G protein-coupled receptor 167; G protein-coupled receptor MrgE; GPR167; G-protein coupled receptor 167; MAS related GPR family member E; mas-related G protein-coupled MRGE; Mas-related G protein-coupled receptor F; MAS-related gene E; MAS-related GPR, member E; mas-related G-protein coupled receptor member E; MAS-related G-protein coupled receptor, member E; MGC138408; Mgrf; MrgE; MRGPRE
Gene Symbols MRGPRE
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human MRGPRE. Recombinant protein control fragment (Product #RP-95167).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116534
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.