Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRGPRF Antibody (916219), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB8396MFV450
Description
MRGPRF Monoclonal antibody specifically detects MRGPRF in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
| MRGPRF | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| GPCR | |
| 116535 | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, CyTOF | |
| 916219 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| FLJ16111, FLJ29034, FLJ40998, FLJ53714, G protein-coupled receptor 140, G protein-coupled receptor 168, G protein-coupled receptor MrgF, GPR140, GPR168, G-protein coupled receptor 140, G-protein coupled receptor 168, mas-related G protein-coupled MRGF, Mas-related gene F protein, MAS-related GPR, member F, mas-related G-protein coupled receptor member F, MGC21621, mrgF, MRGFDKFZp586B2122, RTA, seven transmembrane helix receptor | |
| NS0 mouse myeloma cell line transfected with human MRGPRF, Met1-Ser343, Accession # NP_659452 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction