Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRGPRG Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563629
Description
Immunogen sequence: LIYSGLGRQP GKREPLRSVL RRAL Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 62%.
Orphan receptor. May regulate nociceptor function and/or development, including the sensation or modulation of pain.
Specifications
MRGPRG | |
Polyclonal | |
Unconjugated | |
Mrgprg | |
G protein-coupled receptor 169; G protein-coupled receptor MRGG; GPR169; G-protein coupled receptor 169; MAS related GPR family member G; MAS-related gene G; MAS-related GPR, member G; mas-related G-protein coupled receptor member G; MAS-related G-protein coupled receptor, member G; Mrgg; MRGPRG | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
386746 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86SM5 | |
Mrgprg | |
Recombinant protein corresponding to Human MRGPRG. Recombinant protein control fragment (Product #RP-101534). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction