Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRGPRX1/3/4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987368
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7368 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7368 Supplier Bioss Supplier No. BS6606R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MRGX1 is an opiod receptor that has been reported exclusively in dorsal root ganglion. ESTs for MRGX1 have been identified in a human brain (hypothalamus) library and in a human cancer library.

Specifications

Antigen MRGPRX1/3/4
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Mrgprx1
Gene Accession No. Q96LB2
Gene Alias G protein-coupled receptor MRGX1; G protein-coupled receptor SNSR; G protein-coupled receptor SNSR3; GPCR; MAS related GPR family member X1; Mas-related G protein-coupled receptor G2; MAS-related GPR, member C11; MAS-related GPR, member X1; Mas-related G-protein coupled receptor member C11; Mas-related G-protein coupled receptor member X1; Mgrg2; Mrgc; Mrgc11; Mrgprc; Mrgprc11; MRGPRX1; MRGX1; sensory neuron-specific G protein-coupled receptor 1; Sensory neuron-specific G-protein coupled receptor 1; Sensory neuron-specific G-protein coupled receptor 3/4; Snsr; Snsr1; SNSR3; SNSR4
Gene Symbols Mrgprx1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MRGX1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259249
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.