Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRGPRX4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533955
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533955 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533955 Supplier Invitrogen™ Supplier No. PA533955
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MRGPRX4 Polyclonal Antibody for IHC (P)

Orphan receptor. Probably involved in the function of nociceptive neurons. May regulate nociceptor function and/or development, including the sensation or modulation of pain. Potently activated by enkephalins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRGPRX4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene MRGPRX4
Gene Accession No. Q96LA9
Gene Alias G protein-coupled receptor MRGX4; G protein-coupled receptor SNSR5; G protein-coupled receptor SNSR6; GPCR; MAS related GPR family member X4; MAS-related GPR, member X4; mas-related G-protein coupled receptor member X4; MGC129753; MGC129754; MRGPRX4; MRGX4; sensory neuron-specific G-protein coupled receptor 5/6; SNSR5; SNSR6
Gene Symbols MRGPRX4
Host Species Rabbit
Immunogen Synthetic 12 amino acid peptide from N-terminal extracellular domain of human MRGPRX4 / MRGX4.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117196
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.