Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRGX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579694
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579694 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579694 Supplier Invitrogen™ Supplier No. PA579694
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Marker, Rat Heart Tissue, Rat Skeletal Muscle Tissue, MCF-7 Cell.

MRGX1 is an opiod receptor that has been reported exclusively in dorsal root ganglion. ESTs for MRGX1 have been identified in a human brain (hypothalamus) library and in a human cancer library.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRGX1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene Mrgprx1
Gene Accession No. Q8R4G1
Gene Alias G protein-coupled receptor MRGX1; G protein-coupled receptor SNSR; G protein-coupled receptor SNSR3; GPCR; MAS related GPR family member X1; Mas-related G protein-coupled receptor g2; MAS-related GPR, member C11; MAS-related GPR, member X1; mas-related G-protein coupled receptor member C11; Mas-related G-protein coupled receptor member X1; MGRG2; Mrgc; Mrgc11; Mrgprc; Mrgprc11; mrgprx1; MRGX1; sensory neuron-specific G protein-coupled receptor 1; sensory neuron-specific G-protein coupled receptor 1; Sensory neuron-specific G-protein coupled receptor 3/4; Snsr; Snsr1; SNSR3; SNSR4
Gene Symbols Mrgprx1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the N-terminus of rat MRGX1 (14-28aa TLNKTGHPSCRPILT).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282547
Target Species Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.