Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRGX1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2841700.1ML
Description
MRGX1 Polyclonal specifically detects MRGX1 in Human samples. It is validated for Western Blot.Specifications
MRGX1 | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
G protein-coupled receptor MRGX1, GPCR, MAS-related GPR, member X1, mas-related G-protein coupled receptor member X1, MRGX1G protein-coupled receptor SNSR3, Sensory neuron-specific G-protein coupled receptor 3/4, SNSR3, SNSR4 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human MRGX1 | |
0.1 mL | |
GPCR | |
259249 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction