Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRGX1 Rabbit anti-Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502207819
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-7819 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-7819 Supplier Fabgennix Inc Supplier No. MRGX1101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MRGX1 Polyclonal antibody specifically detects MRGX1 in Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen MRGX1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Mrgprx1
Gene Accession No. Q8CIP3, Q8R4G1
Gene Alias G protein-coupled receptor MRGX1; G protein-coupled receptor SNSR; G protein-coupled receptor SNSR3; GPCR; MAS related GPR family member X1; Mas-related G protein-coupled receptor g2; MAS-related GPR, member C11; MAS-related GPR, member X1; mas-related G-protein coupled receptor member C11; Mas-related G-protein coupled receptor member X1; MGRG2; Mrgc; Mrgc11; Mrgprc; Mrgprc11; mrgprx1; MRGX1; sensory neuron-specific G protein-coupled receptor 1; sensory neuron-specific G-protein coupled receptor 1; Sensory neuron-specific G-protein coupled receptor 3/4; Snsr; Snsr1; SNSR3; SNSR4
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 280-330 on rat MrgX1 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282547, 404242
Target Species Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.