Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRGX2 Antibody (477533), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4727I
Description
MRGX2 Monoclonal antibody specifically detects MRGX2 in Human samples. It is validated for Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOFSpecifications
MRGX2 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOF | |
477533 | |
Flow Cytometry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
MAS-related GPR, member X2, mas-related G-protein coupled receptor member X2, MRGX2G protein-coupled receptor MRGX2 | |
NS0 mouse myeloma cell line transfected with human MRGX2, Met1-Val330, Accession # Q96LB1 | |
0.1 mL | |
GPCR | |
117194 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction