Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRGX2 Antibody (477533), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4727MFV610
Description
MRGX2 Monoclonal antibody specifically detects MRGX2 in Human samples. It is validated for Flow Cytometry, Flow Cytometry, CyTOFSpecifications
| MRGX2 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| GPCR | |
| 117194 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 477533 | |
| Flow Cytometry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| MAS-related GPR, member X2, mas-related G-protein coupled receptor member X2, MRGX2G protein-coupled receptor MRGX2 | |
| NS0 mouse myeloma cell line transfected with human MRGX2, Met1-Val330, Accession # Q96LB1 | |
| RUO | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction