Learn More
Invitrogen™ MRI1 Monoclonal Antibody (OTI9H7)

Description
MRI1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
Specifications
| Antigen | MRI1 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI9H7 |
| Concentration | 1.18 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MRI1 |
| Gene Accession No. | Q9BV20 |
| Gene Alias | 2410018C20Rik; CH73-131E21.1; HAMAP-Rule:MF_03119}; M1Pi; M1Pi {ECO:0000255; mediator of RhoA-dependent invasion; Methylthioribose-1-phosphate isomerase; methylthioribose-1-phosphate isomerase {ECO:0000255; methylthioribose-1-phosphate isomerase 1; methylthioribose-1-phosphate isomerase homolog; methylthioribose-1-phosphate isomerase homolog (S. cerevisiae); MGC3207; MRDI; Mri1; MTNA; MTR-1-P isomerase; MTR-1-P isomerase {ECO:0000255; RGD1307789; similar to CG11334-PB, isoform B; S-methyl-5-thioribose-1-phosphate isomerase; S-methyl-5-thioribose-1-phosphate isomerase {ECO:0000255; S-methyl-5-thioribose-1-phosphate isomerase 1; translation initiation factor eIF-2B subunit alpha/beta/delta-like protein; translation initiation factor eIF-2B subunit alpha/beta/delta-like protein {ECO:0000255; UNQ6390/PRO21135; Ypr118w; zgc:172216 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.