Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRI1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589770
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589770 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589770 Supplier Invitrogen™ Supplier No. PA589770
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTLEAIRYSR GSLQILDQLL LPKQSRYEAV GSVHQAWEAI RAMKVRGAPA IALVGCLSLA VELQAGAGGP GLAALVAFVR DKLSFLVTAR PTAVNMARAA RDLADVAARE AEREGATEEA VRERVICCTE DMLEKDLRDN RSIGDLGARH LLERVAPSGG KVTVLTHCNT GALATAGYGT ALGVIRSLHS LGRLEHAFCT ETRPYNQGAR LTAFELVYEQ IPATLITDSM VAAAMAHRGV SAVVVGADRV VANGDTANKV GTYQLAIVAK HHGIPFYVAA PSSSCDLRLE TGKEIIIEER PGQELTDVNG VRIAAPGIGV WNPAFDVTPH DLITGGIITE LGVFAPEELR TALTTTISSR DGTLDGPQM; Positive Samples: U-87MG, HepG2, HT-1080, Mouse heart, Mouse kidney, Rat brain; Cellular Location: Cell projection, Cytoplasm, Nucleus.

MGC3207 catalyzes the interconversion of methylthioribose-1-phosphate (MTR-1-P) into methylthioribulose-1-phosphate (MTRu-1-P). Independently from catalytic activity, MGC3207 promotes cell invasion in response to constitutive RhoA activation by promoting FAK tyrosine phosphorylation and stress fiber turnover.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRI1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MRI1
Gene Accession No. Q5HZE4, Q9BV20, Q9CQT1
Gene Alias 2410018C20Rik; CH73-131E21.1; HAMAP-Rule:MF_03119}; M1Pi; M1Pi {ECO:0000255; mediator of RhoA-dependent invasion; Methylthioribose-1-phosphate isomerase; methylthioribose-1-phosphate isomerase {ECO:0000255; methylthioribose-1-phosphate isomerase 1; methylthioribose-1-phosphate isomerase homolog; methylthioribose-1-phosphate isomerase homolog (S. cerevisiae); MGC3207; MRDI; Mri1; MTNA; MTR-1-P isomerase; MTR-1-P isomerase {ECO:0000255; RGD1307789; similar to CG11334-PB, isoform B; S-methyl-5-thioribose-1-phosphate isomerase; S-methyl-5-thioribose-1-phosphate isomerase {ECO:0000255; S-methyl-5-thioribose-1-phosphate isomerase 1; translation initiation factor eIF-2B subunit alpha/beta/delta-like protein; translation initiation factor eIF-2B subunit alpha/beta/delta-like protein {ECO:0000255; UNQ6390/PRO21135; Ypr118w; zgc:172216
Gene Symbols MRI1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-369 of human MRI1 (NP_0010268971).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 288912, 67873, 84245
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.