Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MRK 016
GREENER_CHOICE

Catalog No. 381710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-171-0 10 mg
38-175-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 38-171-0 Supplier Tocris Bioscience™ Supplier No. 3817/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

α5-selective GABAA inverse agonist

GABAA receptor inverse agonist selective for the α5-subtype (EC50 = 3 nM). Exhibits affinity at benzodiazepine binding site of recombinant human GABAA receptors (Ki values are 0.77 nM, 0.83 nM, 0.85 nM and 1.4 nM for α3-, α1-, α2-, and α5-containing respectively). Increases long-term potentiation (LTP) in mouse hippocampal slices. Exhibits no anxiogenic or proconvulsant activity.

Chemical Identifiers

CAS 783331-24-8
Molecular Formula C17H20N8O2

Specifications

Quantity 10 mg
Formula Weight 368.39
Purity >98%
Molecular Weight (g/mol) 368.401
Chemical Name or Material MRK 016
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.