Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MRK 560
GREENER_CHOICE

Catalog No. 400010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-001-0 10 mg
40-005-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 40-001-0 Supplier Tocris Bioscience™ Supplier No. 4000/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

γ-secretase inhibitor; attenuates amyloid plaque deposition

γ-secretase inhibitor; inhibits proteolytic cleavage of amyloid precursor protein (APP) over the Notch pathway. Reduces levels of Aβ in the brain (inhibits Aβ40 and Aβ42 in SH-SY5Y neuroblastoma cells with an IC50 of 0.65 nM); attenuates plaque deposition. Orally bioavailable.

Chemical Identifiers

CAS 677772-84-8
Molecular Formula C19H17ClF5NO4S2

Specifications

Quantity 10 mg
Formula Weight 517.92
Purity >99%
Molecular Weight (g/mol) 517.91
Chemical Name or Material MRK 560
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.