Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRLC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546308
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546308 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546308 Supplier Invitrogen™ Supplier No. PA546308
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EKLNGTDPED VIRNAFACFD EEATGTIQED YLRELLTTMG DRFTDEEVDE Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 86%.

MRLC2 is involved in the the activity of nonmuscle myosin II is regulated by phosphorylation of a regulatory light chain. This phosphorylation results in higher MgATPase activity and the assembly of myosin II filaments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRLC2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MYL12B
Gene Accession No. O14950
Gene Alias 1500001M02Rik; C77744; MLC-2; MLC20; MLC-2A; MLCB; MLC-B; Mrlc2; MRLC3; Mrlcb; Myl12b; MYLC2B; myosin light chain 12B; myosin light chain, regulatory B; myosin regulatory light chain 12B; myosin regulatory light chain 2; Myosin regulatory light chain 20 kDa; myosin regulatory light chain 2-B, smooth muscle isoform; myosin regulatory light chain MRCL2; myosin regulatory light chain MRLC2; myosin regulatory light chain-B; myosin regulatory light polypeptide 9-like; myosin RLC-B; myosin, light chain 12B, regulatory; RLC; RLC-B; SHUJUN-1
Gene Symbols MYL12B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human MYL12B (aa 92-141).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103910
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.