Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRM1 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306318MFV610
Description
MRM1 Polyclonal antibody specifically detects MRM1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
MRM1 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
EC 2.1.1.-, FLJ22578, Mitochondrial large ribosomal RNA ribose methylase, mitochondrial rRNA methyltransferase 1 homolog (S. cerevisiae), rRNA methyltransferase 1, mitochondrial | |
Produced in rabbits immunized with E. coli-derived Human MRM1 fragment. | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
79922 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction