Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRM1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | MRM1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
MRM1 Polyclonal specifically detects MRM1 in Human samples. It is validated for Western Blot.Specifications
MRM1 | |
Polyclonal | |
Rabbit | |
Q6IN84 | |
79922 | |
Synthetic peptides corresponding to MRM1(mitochondrial rRNA methyltransferase 1 homolog (S. cerevisiae)) The peptide sequence was selected from the C terminal of MRM1. Peptide sequence LVLGNEGSGLSQEVQASCQLLLTILPRRQLPPGLESLNVSVAAGILLHSI. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
EC 2.1.1.-, FLJ22578, Mitochondrial large ribosomal RNA ribose methylase, mitochondrial rRNA methyltransferase 1 homolog (S. cerevisiae), rRNA methyltransferase 1, mitochondrial | |
MRM1 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title