Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113716
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113716 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113716 Supplier Invitrogen™ Supplier No. PA5113716
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVLGNEGSGL SQEVQASCQL LLTILPRRQL PPGLESLNVS VAAGILLHSI For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 92%.

MRM1 belongs to the RNA methyltransferase trmH family and probably methylates the ribose of guanosine G-2270 in the peptidyl transferase center of the mitochondrial large ribosomal RNA (21S).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRM1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MRM1
Gene Accession No. Q6IN84
Gene Alias 16S rRNA (guanosine(1145)-2'-O)-methyltransferase; 16S rRNA [Gm1145] 2'-O-methyltransferase; A530065E19Rik; CTB-75G16.3; ENSMUSG00000054212; mitochondrial large ribosomal RNA ribose methylase; mitochondrial rRNA methyltransferase 1; mitochondrial rRNA methyltransferase 1 homolog; mitochondrial rRNA methyltransferase 1 homolog (S. cerevisiae); Mrm1; RGD1566232; rRNA methyltransferase 1, mitochondrial
Gene Symbols MRM1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C terminal region of human MRM1 (aa 254-303).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79922
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.