Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MROH1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560720
Description
Immunogen sequence: PGTLPHCAVL HTLASLSVAN AFGVVPFLPS VLSSLLPVLG VAKQDTVRVA FCSALQRFSE GALEYLANLD RAPDPTVRKD AFATDIFSAY DVLFHQWLQS REAKLRL Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 73%.

Specifications
MROH1 | |
Polyclonal | |
Unconjugated | |
MROH1 | |
D330001F17Rik; HEAT repeat containing 7A; HEAT repeat-containing protein 7A; HEATR7A; KIAA1833; maestro heat like repeat family member 1; maestro heat-like repeat family member 1; maestro heat-like repeat-containing protein family member 1; MROH1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
727957 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NDA8 | |
MROH1 | |
Recombinant protein corresponding to Human MROH1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction