Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MROH2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562553
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562553 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562553 Supplier Invitrogen™ Supplier No. PA562553
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TVLNVLHDFE ERIQESEQSW QISAWRKDHP WRRETVKSAL MVMYSCVASY CHPQLLLNLV DSPITAKIIH HYV Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 92%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen MROH2A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MROH2A
Gene Accession No. A6NES4
Gene Alias HEAT repeat containing 7B1; HEAT repeat-containing protein 7B1; HEATR7B1; maestro heat like repeat family member 2A; maestro heat-like repeat family member 2A; maestro heat-like repeat-containing protein family member 2A; MROH2A
Gene Symbols MROH2A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MROH2A. Recombinant protein control fragment (Product #RP-99691).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339766
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.