Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MROH2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563729
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563729 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563729 Supplier Invitrogen™ Supplier No. PA563729
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DKEHIQFLYE RSMDALGKLL KTMMWDNVNA EDCQEMFNLL QMWLVSQKEW ERERAFQITA KVLTNDIEAP ENFKIGSLLG LLAPHSCDTL PTI Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 29%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen MROH2B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MROH2B
Gene Accession No. Q7Z745
Gene Alias HEAT repeat family member 7B2; HEAT repeat family member B2; HEAT repeat-containing protein 7B2; HEATR7B2; maestro heat like repeat family member 2B; maestro heat-like repeat family member 2B; maestro heat-like repeat-containing protein family member 2B; MROH2B; Sperm PKA-interacting factor; SPIF
Gene Symbols MROH2B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MROH2B. Recombinant protein control fragment (Product #RP-100233).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133558
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.