Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MROH7 Monoclonal Antibody (A5-D4-A6)
GREENER_CHOICE

Catalog No. PIMA532885
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532885 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532885 Supplier Invitrogen™ Supplier No. MA532885
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MROH7 Monoclonal Antibody for IHC (P), Western Blot

MROH7 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MROH7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone A5-D4-A6
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% Glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene MROH7
Gene Accession No. Q68CQ1
Gene Alias C1orf175; Gm1027; HEAT repeat containing 8; HEAT repeat-containing protein 8; Heatr8; maestro heat like repeat family member 7; maestro heat-like repeat family member 7; maestro heat-like repeat-containing protein family member 7; MROH7; RGD1563440
Gene Symbols MROH7
Host Species Mouse
Immunogen Recombinant protein within Human C1orf175 aa 1,025-1,323.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374977
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.