Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MROH7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571120
Description
This target displays homology in the following species: Human: 100%.
MROH7 is a protein coding gene.
Specifications
MROH7 | |
Polyclonal | |
Unconjugated | |
MROH7 | |
C1orf175; Gm1027; HEAT repeat containing 8; HEAT repeat-containing protein 8; Heatr8; maestro heat like repeat family member 7; maestro heat-like repeat family member 7; maestro heat-like repeat-containing protein family member 7; MROH7; RGD1563440 | |
Rabbit | |
Affinity chromatography | |
RUO | |
374977 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q68CQ1 | |
MROH7 | |
synthetic peptide directed towards the N-terminal region of human HEATR8. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction