Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRP1 Antibody (IU2H10), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB400156B
Description
MRP1 Monoclonal specifically detects MRP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
MRP1 | |
Monoclonal | |
Biotin | |
P33527 | |
ABCC1 | |
Recombinant peptide (a.a. 1-33) of human MRP1. [UniProt# P33527]. | |
0.1 mL | |
ABC Transporters, Amino Acids Drugs and other small molecules, Cancer, Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction | |
4363 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
IU2H10 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
ABC29, ABCC, ATP-binding cassette sub-family C member 1, ATP-binding cassette, sub-family C (CFTR/MRP), member 1, EC 3.6.3, EC 3.6.3.44, GS-X, Leukotriene C(4) transporter, LTC4 transporter, MRP1DKFZp781G125, MRPDKFZp686N04233, multidrug resistance associated protein 1, multidrug resistance protein, multidrug resistance-associated protein 1, multiple drug resistance protein 1, multiple drug resistance-associated protein | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction