Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRP1 Antibody (MRPm6), Novus Biologicals™
SDP

Catalog No. p-7161267 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP14234025 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP14234025 Supplier Novus Biologicals Supplier No. NBP1423400.25ML

Mouse Monoclonal Antibody

MRP1 Monoclonal specifically detects MRP1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen MRP1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MRPm6
Conjugate Unconjugated
Dilution Western Blot 1:20-1:50, Flow Cytometry 1:20-1:50, Immunohistochemistry 1:20-50, Immunohistochemistry-Paraffin 1:20-50, Immunohistochemistry-Frozen 1:20-50
Formulation Tissue culture supernatant with 0.1% sodium azide
Gene Alias ABC29, ABCC, ATP-binding cassette sub-family C member 1, ATP-binding cassette, sub-family C (CFTR/MRP), member 1, EC 3.6.3, EC 3.6.3.44, GS-X, Leukotriene C(4) transporter, LTC4 transporter, MRP1DKFZp781G125, MRPDKFZp686N04233, multidrug resistance associated protein 1, multidrug resistance protein, multidrug resistance-associated protein 1, multiple drug resistance protein 1, multiple drug resistance-associated protein
Gene Symbols ABCC1
Host Species Mouse
Immunogen The antibody reacts with the MRP1 antigen.
Quantity 0.25 mL
Primary or Secondary Primary
Gene ID (Entrez) 4363
Test Specificity MRPm6 reacts with an internal epitope of MRP, a 180-195 kD transmembrane transporter protein overexpressed in various human non-P-glycoprotein MDR tumor cell lines. MRPm6 was raised against a bacterial fusion protein of MRP, containing a segment of 170 amino acids in the carboxy terminal end and part of the carboxy proximal nucleotide binding domain of the protein. MRPm6 does not cross react with the human MDR1 and MDR3 gene products (Flens et al. 1994)
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.