Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578694
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578694 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578694 Supplier Invitrogen™ Supplier No. PA578694
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: JURKAT Cell, CEM Cell, A549 Cell. IHC: Human Lung Cancer Tissue.

The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra-and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This full transporter is a member of the MRP subfamily which is involved in multi-drug resistance. This protein functions as a multispecific organic anion transporter, with oxidized glutathione, cysteinyl leukotrienes, and activated aflatoxin B1 as substrates. This protein also transports glucuronides and sulfate conjugates of steroid hormones and bile salts. Alternative splicing by exon deletion results in several splice variants but maintains the original open reading frame in all forms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene ABCC1
Gene Accession No. O35379, P33527, Q8CG09
Gene Alias ABC29; ABCC; Abcc1; Abcc1a; Abcc1b; ATP binding cassette subfamily C member 1; ATP-binding cassette sub-family C ( CFTR/MRP) member 1a; ATP-binding cassette sub-family C member 1; ATP-binding cassette transporter variant ABCC1delta-ex13; ATP-binding cassette transporter variant ABCC1delta-ex13 ATP-binding cassette transporter variant ABCC1delta-ex25; ATP-binding cassette transporter variant ABCC1delta-ex25&26; ATP-binding cassette, subfamily C (CFTR/MRP), member 1; ATP-binding cassette, sub-family C (CFTR/MRP), member 1; ATP-binding cassette, sub-family C (CFTR/MRP), member 1a; ATP-binding cassette, sub-family C (CFTR/MRP), member 1b; Avcc1a; DKFZp686N04233; DKFZp781G125; Glutathione-S-conjugate-translocating ATPase ABCC1; GS-X; leuk; leukotriene C(4) transporter; LTC4 transporter; Mdrap; MRP; Mrp1; multidrug resistance protein 1; multidrug resistance-associated protein 1; multiple drug resistance-associated protein
Gene Symbols ABCC1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human MRP1 (1514-1531aa, LLQQRGLFYSMAKDAGLV).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17250, 24565, 4363
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.