Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582015
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582015 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582015 Supplier Invitrogen™ Supplier No. PA582015
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVTHSMSYLP QVDVIIVMSG GKISEMGSYQ ELLARDGAFA EFLRTYASTE QEQDAEENGV TGVSGPGKEA KQMENGMLVT DSAGKQLQRQ LSSSSSYSGD ISRHHNSTAE LQKAEAKKEE TWKLMEADKA QTGQVKLSVY WD Highest antigen sequence indentity to the following orthologs: Mouse - 76%, Rat - 73%.

The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra-and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This full transporter is a member of the MRP subfamily which is involved in multi-drug resistance. This protein functions as a multispecific organic anion transporter, with oxidized glutathione, cysteinyl leukotrienes, and activated aflatoxin B1 as substrates. This protein also transports glucuronides and sulfate conjugates of steroid hormones and bile salts. Alternative splicing by exon deletion results in several splice variants but maintains the original open reading frame in all forms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ABCC1
Gene Accession No. P33527
Gene Alias ABC29; ABCC; Abcc1; Abcc1a; Abcc1b; ATP binding cassette subfamily C member 1; ATP-binding cassette sub-family C ( CFTR/MRP) member 1a; ATP-binding cassette sub-family C member 1; ATP-binding cassette transporter variant ABCC1delta-ex13; ATP-binding cassette transporter variant ABCC1delta-ex13&14; ATP-binding cassette transporter variant ABCC1delta-ex25; ATP-binding cassette transporter variant ABCC1delta-ex25&26; ATP-binding cassette, subfamily C (CFTR/MRP), member 1; ATP-binding cassette, sub-family C (CFTR/MRP), member 1; ATP-binding cassette, sub-family C (CFTR/MRP), member 1a; ATP-binding cassette, sub-family C (CFTR/MRP), member 1b; Avcc1a; DKFZp686N04233; DKFZp781G125; Glutathione-S-conjugate-translocating ATPase ABCC1; GS-X; leuk; leukotriene C(4) transporter; LTC4 transporter; Mdrap; MRP; Mrp1; multidrug resistance protein 1; multidrug resistance-associated protein 1; multiple drug resistance-associated protein
Gene Symbols ABCC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ABCC1. Recombinant protein control fragment (Product #RP-101755).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4363
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.