Learn More
Invitrogen™ MRP7 Polyclonal Antibody

Description
The antibody detects endogenous levels of total ABCC10 protein.
Specifications
Specifications
| Antigen | MRP7 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | ABCC10 |
| Gene Accession No. | Q5T3U5 |
| Gene Alias | ABCC10; ATP binding cassette subfamily C member 10; ATP-binding cassette sub-family C member 10; ATP-binding cassette, subfamily C (CFTR/MRP), member 10; ATP-binding cassette, sub-family C (CFTR/MRP), member 10; EST182763; mFLJ00002; Mrp7; Multidrug resistance-associated protein 7; SIMRP7 |
| Gene Symbols | ABCC10 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.