Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRP2 Monoclonal Antibody (M2I-4)
GREENER_CHOICE

Catalog No. PIMA126535
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA126535 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA126535 Supplier Invitrogen™ Supplier No. MA126535
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-26535 detects multidrug resistance associated protein 2 from human samples.

This gene encodes the prototype of the membrane-associated guanylate kinase (MAGUK) family proteins. MAGUKs interact with the cytoskeleton and regulate cell proliferation, signaling pathways, and intercellular junctions. The encoded protein is an extensively palmitoylated membrane phosphoprotein containing a PDZ domain, a Src homology 3 (SH3) motif, and a guanylate kinase domain. This gene product interacts with various cytoskeletal proteins and cell junctional proteins in different tissue and cell types, and may be involved in the regulation of cell shape, hair cell development, neural patterning of the retina, and apico-basal polarity and tumor suppression pathways in non-erythroid cells. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRP2
Applications Flow Cytometry, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone M2I-4
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.1% sodium azide
Gene ABCC2
Gene Accession No. Q92887
Gene Alias ABC30; Abcc2; AI173996; ATP binding cassette subfamily C member 2; ATP-binding cassette sub-family C member 2; ATP-binding cassette, subfamily C (CFTR/MRP), member 2; ATP-binding cassette, sub-family C (CFTR/MRP), member 2; calicular multispecific organic anion transporter; canalicular multidrug resistance protein; Canalicular multispecific organic anion transporter 1; Cmoat; CMOAT1; Cmrp; DJS; Human MRP2; KIAA1010; MRP2; multidrug resistance associated protein 2; multidrug resistance protein 2; multidrug resistance-associated protein 2
Gene Symbols ABCC2
Host Species Mouse
Immunogen Bacterial fusion protein of human MRP 2 containing amino acids of 215-310.
Purification Method Purified
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1244
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.