Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ MRP2, Mouse, Unlabeled, Clone: 4C9.2,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABN1545
Description
Canalicular multispecific organic anion transporter 1 (UniProt Q92887; also known as ATP-binding cassette sub-family C member 2, Canalicular multidrug resistance protein, cMRP, Multidrug resistance-associated protein 2) is encoded by the ABCC2 (also known as CMOAT, CMOAT1, CMRP, DJS, MRP2) gene (Gene ID 1244) in human. MRP2/ABCC2 belongs to the family of human ATP-binding cassette (ABC) transporters that are classified into seven subfamilies from ABC-A to ABC-G based on their sequence similarities. P-glycoprotein (P-gp/ABCB1), MRP2/ABCC2, and breast cancer resistance protein (BCRP/ABCG2) are well known plasma membrane ABC transporters that mediate the export of intracellular drugs, drug conjugates and metabolites. MRP2/ABCC2 is the second member of the ABCC subfamily of 13 transporters (ABCC1 to ABCC13). MRP2 is expressed in the apical hepatocyte plasma membrane, renal proximal tubules and small intestine, where it plays a role in the elimination and oral bioavailability of drugs, xenotoxins and their metabolites. MRP2 primarily functions as an organic anion transporter to mediate the elimination of bile acids, GSH and conjugated metabolites of numerous drugs and other xenobiotics. Weakly basic drugs appear to be co-transported with GSH by MRP2. Methotrexate, anthracyclines (doxorubicin, epirubicin), mitoxantrone, cisplatin, and etoposide are well known chemotherapeutic substrates of MRP2. MRP2 contains 17-transmembrane helices (a.a. 28-48, 69-89, 94-114, 127-147, 166-186, 314-334, 361-381, 438-458, 462-482, 545-565, 588-608, 972-992, 1034-1054, 1098-1118, 1120-1140, 1212-1232, 1235-1255), having its N-terminal end (a.a. 1-27) exposed extracellularly and C-terminal end (a.a. 1256-1545) at the cytoplasmic side. Two conserved nucleotide-binding domains (NBDs; a.a. 671-678 and 1334-1341) mediate the binding and hydrolyzing ATP via an ATPase, thereby providing energy for substrates export from the cytoplasm to the extracellular space.
Specifications
MRP2 | |
Monoclonal | |
Please refer to lot specific datasheet. | |
Purified mouse IgG2b in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide. | |
Q92887 | |
GST-tagged recombinant fragment corresponding to the third cytoplasmic domain of human MRP2. | |
100 μg | |
Neuroscience | |
NP_000383 | |
Human | |
Purified |
Western Blot | |
4C9.2 | |
Unconjugated | |
ABCC2, CMOAT, CMOAT1, CMRP, DJS, MRP2 | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Clone 4C9.2 targets an epitope within the third cytoplasmic domain of human MRP2. | |
Stable for 1 year at 2°-8°C from date of receipt. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction