Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585631
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585631 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585631 Supplier Invitrogen™ Supplier No. PA585631
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Rhesus Monkey (100%). Positive Control: A549, A549 membrane extract. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes the prototype of the membrane-associated guanylate kinase (MAGUK) family proteins. MAGUKs interact with the cytoskeleton and regulate cell proliferation, signaling pathways, and intercellular junctions. The encoded protein is an extensively palmitoylated membrane phosphoprotein containing a PDZ domain, a Src homology 3 (SH3) motif, and a guanylate kinase domain. This gene product interacts with various cytoskeletal proteins and cell junctional proteins in different tissue and cell types, and may be involved in the regulation of cell shape, hair cell development, neural patterning of the retina, and apico-basal polarity and tumor suppression pathways in non-erythroid cells. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRP2
Applications Western Blot
Classification Polyclonal
Concentration 1.76 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene ABCC2
Gene Accession No. Q92887
Gene Alias ABC30; Abcc2; AI173996; ATP binding cassette subfamily C member 2; ATP-binding cassette sub-family C member 2; ATP-binding cassette, subfamily C (CFTR/MRP), member 2; ATP-binding cassette, sub-family C (CFTR/MRP), member 2; calicular multispecific organic anion transporter; canalicular multidrug resistance protein; Canalicular multispecific organic anion transporter 1; Cmoat; CMOAT1; Cmrp; DJS; Human MRP2; KIAA1010; MRP2; multidrug resistance associated protein 2; multidrug resistance protein 2; multidrug resistance-associated protein 2
Gene Symbols ABCC2
Host Species Rabbit
Immunogen Synthetic peptide encompassing a sequence within the Intracellular domain of human MRP2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1244
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.