Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRP3 Antibody (ABCC3/2971), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279837G
Description
MRP3 Monoclonal specifically detects MRP3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MRP3 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ABCC3 | |
Recombinant fragment (around aa 815-957) of human MRP3 protein (exact sequence is proprietary) (Uniprot: O15438) | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
ABCC3/2971 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
ABC31, ATP-binding cassette sub-family C member 3, ATP-binding cassette, sub-family C (CFTR/MRP), member 3, canalicular multispecific organic anion transporter 2, canicular multispecific organic anion transporter, cMOAT2, EC 3.6.3, EC 3.6.3.44, EC 6.3.2.4, MLP2DKFZp686E22157, MRP3MOAT-DEST90757, multidrug resistance associated protein, Multidrug resistance-associated protein 3, Multi-specific organic anion transporter D | |
Mouse | |
Protein A or G purified | |
RUO | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction