Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRP4/ABCC4 Antibody (M4I-80), Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP142338
Description
MRP4/ABCC4 Monoclonal specifically detects MRP4/ABCC4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
MRP4/ABCC4 | |
Monoclonal | |
Unconjugated | |
ATP-binding cassette sub-family C member 4, ATP-binding cassette, sub-family C (CFTR/MRP), member 4, EST170205, MOAT-BbA464I2.1 (ATP-binding cassette, sub-family C (CFTR/MRP), member 4), MOATBcanalicular multispecific organic anion transporter (ABC superfamily), MRP4MRP/cMOAT-related ABC transporter, multidrug resistance-associated protein 4, multispecific organic anion transporter B, Multi-specific organic anion transporter B | |
Rat | |
Tissue culture supernatant | |
RUO | |
Primary | |
The M4I-80 Mab was selected after immunization with a fusion protein containing the E. coli maltose binding protein and a fragment of the human MRP4 protein corresponding to amino acids 372-431. MRP4 transports cyclic nucleotides and anti-retroviral compounds. The M4I-80 Mab also reacts with the mouse orthologue of the transporter molecule (Mrp4). | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
M4I-80 | |
Western Blot 1:20-1:50, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:20-1:50, Immunohistochemistry-Frozen 1:20 | |
ABCC4 | |
The antibody reacts with the MRP4 antigen. | |
0.2 mL | |
Lipid and Metabolism | |
10257 | |
Human, Mouse | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction