Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRP4/ABCC4 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 9 publications
Supplier: Novus Biologicals NB1001471
Description
MRP4/ABCC4 Polyclonal specifically detects MRP4/ABCC4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Peptide ELISA.Specifications
MRP4/ABCC4 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
ATP-binding cassette sub-family C member 4, ATP-binding cassette, sub-family C (CFTR/MRP), member 4, EST170205, MOAT-BbA464I2.1 (ATP-binding cassette, sub-family C (CFTR/MRP), member 4), MOATBcanalicular multispecific organic anion transporter (ABC superfamily), MRP4MRP/cMOAT-related ABC transporter, multidrug resistance-associated protein 4, multispecific organic anion transporter B, Multi-specific organic anion transporter B | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is expected to recognize isoform 1 (NP_005836.2) only. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), ELISA | |
0.5 mg/mL | |
Western Blot 1-3 μg/mL, Immunohistochemistry 4 - 6 μg/mL, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 4 - 6 μg/mL, Immunohistochemistry-Frozen, Peptide ELISA Detection limit 1:1000 | |
O15439 | |
ABCC4 | |
Peptide with sequence CNGQPSTLTIFETAL corresponding to C-Terminus according to NP_005836.2. | |
0.1 mg | |
Lipid and Metabolism | |
10257 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction