Learn More
Invitrogen™ MRP4 Monoclonal Antibody (M4I-80)

Description
The M4I-80 Mab was selected after immunization with a fusion protein containing the E. coli maltose binding protein and a fragment of the human MRP4 protein corresponding to amino acids 372-431. MRP4 transports cyclic nucleotides and anti-retroviral compounds. The M4I-80 Mab also reacts with the mouse orthologue of the transporter molecule (Mrp4).
Specifications
Specifications
| Antigen | MRP4 |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | M4I-80 |
| Concentration | 200 μg/mL |
| Conjugate | Unconjugated |
| Formulation | tissue culture supernatant with 0.7% BSA and 0.1% sodium azide |
| Gene | ABCC4 |
| Gene Accession No. | O15439 |
| Gene Alias | ABCC4; ABC-tranporter; ATP binding cassette subfamily C member 4; ATP-binding cassette protein C4; ATP-binding cassette sub-family C member 4; ATP-binding cassette, subfamily C (CFTR/MRP), member 4; ATP-binding cassette, sub-family C (CFTR/MRP), member 4; bA464I2.1 (ATP-binding cassette, sub-family C (CFTR/MRP), member 4); canalicular multispecific organic anion transporter (ABC superfamily); D630049P08Rik; EST170205; MOATB; MOAT-B; MRP/cMOAT-related ABC transporter; MRP4; Multidrug resistance-associated protein 4; multi-specific organic anion transporter B; RP11-74A12.1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.