Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578695
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578695 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578695 Supplier Invitrogen™ Supplier No. PA578695
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Human ATF1 shares 91% amino acid (aa) sequence identity with mouse ATF1. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - IHC: rat intestine tissue, human intestinal cancer tissue, mouse intestine tissue.

The protein encoded by this gene is a member of the superfamily of ATP-binding cassette transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies. This protein is a member of the MRP subfamily which is involved in multi-drug resistance. The specific function of this protein has not yet been determined; however, this protein may play a role in cellular detoxification as a pump for its substrate, organic anions. Alternative splicing results in multiple splice variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRP4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene ABCC4
Gene Accession No. O15439
Gene Alias ABCC4; ABC-tranporter; ATP binding cassette subfamily C member 4; ATP-binding cassette protein C4; ATP-binding cassette sub-family C member 4; ATP-binding cassette, subfamily C (CFTR/MRP), member 4; ATP-binding cassette, sub-family C (CFTR/MRP), member 4; bA464I2.1 (ATP-binding cassette, sub-family C (CFTR/MRP), member 4); canalicular multispecific organic anion transporter (ABC superfamily); D630049P08Rik; EST170205; MOATB; MOAT-B; MRP/cMOAT-related ABC transporter; MRP4; Multidrug resistance-associated protein 4; multi-specific organic anion transporter B; RP11-74A12.1
Gene Symbols ABCC4
Host Species Rabbit
Immunogen E.coli-derived human MRP4 recombinant protein (Position: M1-P370).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10257, 170924, 239273
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.