Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550520
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550520 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550520 Supplier Invitrogen™ Supplier No. PA550520
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total ABCC5 protein.

The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MRP subfamily which is involved in multi-drug resistance. This protein functions in the cellular export of its substrate, cyclic nucleotides. This export contributes to the degradation of phosphodiesterases and possibly an elimination pathway for cyclic nucleotides. Studies show that this protein provides resistance to thiopurine anticancer drugs, 6-mercatopurine and thioguanine, and the anti-HIV drug 9-(2-phosphonylmethoxyethyl)adenine. This protein may be involved in resistance to thiopurines in acute lymphoblastic leukemia and antiretroviral nucleoside analogs in HIV-infected patients. Alternative splicing of this gene has been detected; however, the complete sequence and translation initiation site is unclear.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRP5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene ABCC5
Gene Accession No. O15440, Q9R1X5
Gene Alias 2900011L11Rik; ABC 33; ABC33; ABCC 5; ABCC5; Abcc5a; Abcc5b; AI132311; ATP binding cassette subfamily C member 5; ATP-binding cassette sub-family C (CFTR/MRP) member 5a; ATP-binding cassette sub-family C member 5; ATP-binding cassette, subfamily C (CFTR/MRP), member 5; ATP-binding cassette, sub-family C (CFTR/MRP), member 5; ATP-binding cassette, sub-family C (CFTR/MRP), member 5a; ATP-binding cassette, sub-family C (CFTR/MRP), member 5b; ATP-binding cassette, sub-family C, member 5; canalicular multispecific organic anion transporter C; DKFZp686C1782; EST277145; MOAT C; MOATC; MOAT-C; MRP 5; MRP5; multidrug resistance-associated protein 5; Multi-specific organic anion transporter C; pABC 11; pABC11; SMRP
Gene Symbols ABCC5
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues near the N terminal of human ATP-binding cassette, sub-family C.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10057, 27416
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.