Learn More
Invitrogen™ MRP5 Polyclonal Antibody

Description
Immunogen sequence: MKDIDIGKEY IIPSPGYRSV RERTSTSGTH RDREDSKFRR TRPLECQDAL ETAARAEGLS LDASMHSQLR ILDEEHPKGK YHHGLSALKP IRTTSKHQHP VDNAGLFSCM TFSWLSSLAR VAHKKGELSM EDVWSLSKHE SSDVNCRRLE RLWQEELNEV GPDAASLRRV VWIFCRTRL; Positive Samples: Mouse brain, Mouse lung, Rat brain; Cellular Location: Membrane, Multi-pass membrane protein.
Specifications
Specifications
| Antigen | MRP5 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.58 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ABCC5 |
| Gene Accession No. | O15440, Q9QYM0, Q9R1X5 |
| Gene Alias | 2900011L11Rik; ABC 33; ABC33; ABCC 5; ABCC5; Abcc5a; Abcc5b; AI132311; ATP binding cassette subfamily C member 5; ATP-binding cassette sub-family C (CFTR/MRP) member 5a; ATP-binding cassette sub-family C member 5; ATP-binding cassette, subfamily C (CFTR/MRP), member 5; ATP-binding cassette, sub-family C (CFTR/MRP), member 5; ATP-binding cassette, sub-family C (CFTR/MRP), member 5a; ATP-binding cassette, sub-family C (CFTR/MRP), member 5b; ATP-binding cassette, sub-family C, member 5; canalicular multispecific organic anion transporter C; DKFZp686C1782; EST277145; MOAT C; MOATC; MOAT-C; MRP 5; MRP5; multidrug resistance-associated protein 5; Multi-specific organic anion transporter C; pABC 11; pABC11; SMRP |
| Gene Symbols | ABCC5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.