Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118970
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118970 200 μL
PIPA5122298 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5118970 Supplier Invitrogen™ Supplier No. PA5118970
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MRP6 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). The encoded protein, a member of the MRP subfamily, is involved in multi-drug resistance. Mutations in this gene cause pseudoxanthoma elasticum. Alternatively spliced transcript variants that encode different proteins have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRP6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene ABCC6
Gene Accession No. O88269
Gene Alias ABC34; Abcc1b; ABCC6; Anthracycline resistance-associated protein; ARA; ATP binding cassette subfamily C member 6; ATP-binding cassette sub-family C member 6; ATP-binding cassette, subfamily C (CFTR/MRP), member 6; ATP-binding cassette, sub-family C (CFTR/MRP), member 6; ATP-binding cassette, sub-family C, member 6; DCC; dyscalc; Dyscalc1; EST349056; GACI2; liver multidrug resistance-associated protein 6; Mlp1; MLP-1; MOATE; MOAT-E; Mrp6; MRP-like protein 1; Multidrug resistance-associated protein 6; multidrug resistance-associated protein-6; multi-specific organic anion transporter E; PXE; PXE1; URG7; URG7 protein; multidrug resistance-associated protein 6
Gene Symbols ABCC6
Host Species Rabbit
Immunogen Recombinant protein ATP Binding Cassette Transporter C6. The antigen corresponds to amino acid range 1260-1502 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81642
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.