Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCC11 Monoclonal Antibody (2C5/4)
GREENER_CHOICE

Catalog No. PIMA126154
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA126154 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA126154 Supplier Invitrogen™ Supplier No. MA126154
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-26154 detects MRP8 from human samples.

ABCC11 is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This ABC full transporter is a member of the MRP subfamily which is involved in multi-drug resistance. The product of this gene participates in physiological processes involving bile acids, conjugated steroids, and cyclic nucleotides. In addition, a SNP in this gene is responsible for determination of human earwax type. This gene and family member ABCC12 are determined to be derived by duplication and are both localized to chromosome 16q12.1. Multiple alternatively spliced transcript variants have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCC11
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2C5/4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ABCC11
Gene Accession No. Q96J66
Gene Alias ABCC11; ATP binding cassette subfamily C member 11; ATP-binding cassette protein C11; ATP-binding cassette sub-family C member 11; ATP-binding cassette transporter MRP8; ATP-binding cassette transporter sub-family C member 11; ATP-binding cassette, sub-family C (CFTR/MRP), member 11; EWWD; MRP8; multidrug resistance-associated protein 8; multi-resistance protein 8; WW
Gene Symbols ABCC11
Host Species Mouse
Purification Method IgG Fraction
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85320
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.