Learn More
Invitrogen™ MRP9 Polyclonal Antibody

Description
The antibody detects endogenous levels of total ABCC12 protein.
Specifications
Specifications
| Antigen | MRP9 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | ABCC12 |
| Gene Accession No. | Q96J65 |
| Gene Alias | 4930467B22Rik; Abcc12; ATP binding cassette subfamily C member 12; ATP-binding cassette protein C12 variant A; ATP-binding cassette sub-family C member 12; ATP-binding cassette transporter sub-family C member 12; ATP-binding cassette, subfamily C (CFTR/MRP), member 12; ATP-binding cassette, sub-family C (CFTR/MRP), member 12; MRP9; Multidrug resistance-associated protein 9 |
| Gene Symbols | ABCC12 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.