Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPL1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5121765

 View more versions of this product

Catalog No. PIPA5121765


Only null left
Add to Cart
This item is not returnable. View return policy

Description

Description

MRPL1 Polyclonal Antibody for Western Blot, IHC (P)

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein that belongs to the L1 ribosomal protein family.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MRPL1
Polyclonal
Unconjugated
MRPL1
2410002L03Rik; 39S ribosomal protein L1, mitochondrial; 5830418D04Rik; AI462572; BM022; BM-022; L1mt; Mitochondrial large ribosomal subunit protein uL1m; mitochondrial ribosomal protein L1; Mrpl1; MRP-L1
Rabbit
Antigen affinity chromatography, Protein A
RUO
65008
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q9BYD6
MRPL1
Recombinant protein Mitochondrial Ribosomal Protein L1. The antigen corresponds to amino acid range 147-285 of the target protein.
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.