Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPL10 Antibody (OTI2B4), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272779UV
Description
MRPL10 Monoclonal specifically detects MRPL10 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MRPL10 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MRPL10 | |
Full length recombinant protein of human MRPL10 (NP_660298) produced in E.coli. | |
0.1 mL | |
124995.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI2B4 | |
Western Blot, Immunohistochemistry | |
L10mt, MGC17973, mitochondrial ribosomal protein L10, MRP-L10L8mt, MRP-L839S ribosomal protein L10, mitochondrial, MRPL839S ribosomal protein L8, mitochondrial, RPML8L10MT | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction