Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPL10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554357
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554357 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554357 Supplier Invitrogen™ Supplier No. PA554357
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NVALSAEDKL LMRHQLRKHK ILMKVFPNQV LKPFLEDSKY QNLLPLFVGH NMLLVSEEPK VKEMVRILRT VPFLPLLGGC Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 91%.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. A pseudogene corresponding to this gene is found on chromosome 5q.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL10
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MRPL10
Gene Accession No. Q7Z7H8
Gene Alias 0610040E02Rik; 39S ribosomal protein L10, mitochondrial; 39S ribosomal protein L8, mitochondrial; C78440; L10MT; L8mt; Mitochondrial large ribosomal subunit protein uL10m; mitochondrial ribosomal protein L10; MRPL10; MRP-L10; MRPL8; MRP-L8; Rpml8
Gene Symbols MRPL10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MRPL10. Recombinant protein control fragment (Product #RP-93341).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124995
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.