Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPL12 Polyclonal Antibody
MRPL12 Polyclonal Antibody
Supplier: Thermo Scientific PA575535
Description
MRPL12 Polyclonal Antibody for Western Blot

Specifications
MRPL12 | |
Polyclonal | |
Unconjugated | |
MRPL12 | |
0610034O11Rik; 1500031N16Rik; 39S ribosomal protein L12, mitochondrial; 5c5-2; FLJ60124; hypothetical protein LOC550391; im:6893415; L12mt; MGC8610; Mitochondrial large ribosomal subunit protein bL12m; mitochondrial ribosomal protein L12; mitochondrial ribosomal protein L7/12; MRPL12; MRP-L12; MRP-L31/34; MRPL7; MRPL7/L12; ribosomal protein, mitochondrial, L12; Rpm12; RPML12; zgc:112445 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
303746, 56282, 6182 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide; pH 7.2 | |
P52815, Q9DB15 | |
MRPL12 | |
Synthetic peptide corresponding to amino acids 75-120 of Human MRPL12. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction